CAS RN 307-70-0
IUPAC Nor:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol
Synonymes:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol
Poids moléculaire (g/mol):
532.12
Formule moléculaire:
C11H4F20O
InChi Key:
GJYLSFCFHFUODO-UHFFFAOYSA-N
SMILES:
OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F
1
–
1
de
1
résultats
1
1H,1H,11H-Eicosafluoro-1-undecanol 90.0+%, TCI America™
CAS: 307-70-0 Formule moléculaire: C11H4F20O Poids moléculaire (g/mol): 532.12 Numéro MDL: MFCD00039628 Clé InChI: GJYLSFCFHFUODO-UHFFFAOYSA-N Synonyme: 1h,1h,11h-eicosafluoro-1-undecanol,1-undecanol, 1h,1h,11h-eicosafluoro,1,1,11-trihydroperfluoroundecanol,1h,1h,11h-perfluoro-1-undecanol,1h,1h,11h-eicosafluoroundecanol-1,1h,1h,11h-eicosafluoro-1-undecanol, tech.,1h,11h-eicosafluoro-1-undecanol,1h,1h,11h-perfluoroundecan-1-ol,1-undecanol,1h,11h-eicosafluoro,1h, 1h, 11h-eicosafluoroundecanol CID PubChem: 67548 Nom IUPAC: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-icosafluoroundecan-1-ol SMILES: C(C(C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O