accessibility menu, dialog, popup

UserName

CAS RN 306960-67-8

H3COONNH3CFFF

CAS RN 306960-67-8

IUPAC Name: ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
Synonyms: ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
Molecular Weight (g/mol): 234.18
Molecular Formula: C9H9F3N2O2
InChi Key: LYEKBQSEKSEXLT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results