accessibility menu, dialog, popup

UserName

CAS RN 306934-80-5

IUPAC Name: 6-oxo-1,6-dihydropyridazine-3-carboxylic acid hydrate
Synonyms: 6-oxo-1H-pyridazine-3-carboxylic acid hydrate
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: DZCBVXKRERSHAA-UHFFFAOYSA-N
SMILES: O.OC(=O)C1=NNC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results