accessibility menu, dialog, popup

UserName

CAS RN 306-23-0

IUPAC Name: 2-hydroxy-3-(4-hydroxyphenyl)propanoic acid
Synonyms: hydroxyphenyllactic acid hydroxyphenyllactate DL-p-hydroxyphenyllactate para-hydroxyphenyllactic acid p-hydroxyphenyl lactic acid DL-p-hydroxyphenyllactic acid HPLA a,4-dihydroxybenzenepropanoate (p-hydroxyphenyl)-lactic acid 4-hydroxyphenyllactic acid
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: JVGVDSSUAVXRDY-UHFFFAOYNA-N
SMILES: OC(CC1=CC=C(O)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results