accessibility menu, dialog, popup

UserName

CAS RN 3056-17-5

IUPAC Name: 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: stavudine
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: XNKLLVCARDGLGL-HGXVMFPFNA-N
SMILES: CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results