CAS RN 3038-48-0
IUPAC Name:
2-[2-(trifluoromethyl)phenyl]acetate
Synonyms:
2-[2-(trifluoromethyl)phenyl]acetate
Molecular Weight (g/mol):
203.14
Molecular Formula:
C9H6F3O2
InChi Key:
TYOCDHCKTWANIR-UHFFFAOYSA-M
SMILES:
[O-]C(=O)CC1=CC=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™
CAS: 3038-48-0 Molecular Formula: C9H6F3O2 Molecular Weight (g/mol): 203.14 MDL Number: MFCD00004327 InChI Key: TYOCDHCKTWANIR-UHFFFAOYSA-M Synonym: 2-trifluoromethyl phenylacetic acid,2-2-trifluoromethyl phenyl acetic acid,2-trifluoromethylphenylacetic acid,a,a,a-trifluoro-o-tolyl acetic acid,2-trifluoromethyl phenyl acetic acid,alpha,alpha,alpha-trifluoro-o-tolyl acetic acid,benzeneacetic acid, 2-trifluoromethyl,acmc-209hfg,ksc494k8b PubChem CID: 594597 IUPAC Name: 2-[2-(trifluoromethyl)phenyl]acetate SMILES: [O-]C(=O)CC1=CC=CC=C1C(F)(F)F