missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylacetic Acid 98.0+%, TCI America™

Catalog No. T16615G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16615G 5 g
T16611G 1 g
T166125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T16615G Supplier TCI America Supplier No. T16615G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3038-48-0
Molecular Formula C9H6F3O2
Molecular Weight (g/mol) 203.14
MDL Number MFCD00004327
InChI Key TYOCDHCKTWANIR-UHFFFAOYSA-M
Synonym 2-trifluoromethyl phenylacetic acid, 2-2-trifluoromethyl phenyl acetic acid, 2-trifluoromethylphenylacetic acid, a,a,a-trifluoro-o-tolyl acetic acid, 2-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-o-tolyl acetic acid, benzeneacetic acid, 2-trifluoromethyl, acmc-209hfg, ksc494k8b
PubChem CID 594597
IUPAC Name 2-[2-(trifluoromethyl)phenyl]acetate
SMILES [O-]C(=O)CC1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 5 g
Formula Weight 204.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)phenylacetic Acid