CAS RN 30162-37-9
IUPAC Name:
(pyridin-3-yl)thiourea
Synonyms:
pyridin-3-ylthiourea
Molecular Weight (g/mol):
153.2
Molecular Formula:
C6H7N3S
InChi Key:
CFOJQUGXHMGMOT-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=CC=CN=C1
1
–
1
of
1
results
1
Promotions Available
N-(3-Pyridyl)thiourea, 98+%
N-(3-Pyridyl)thiourea, 98+%
CAS: 30162-37-9 Molecular Formula: C6H7N3S Molecular Weight (g/mol): 153.20 MDL Number: MFCD00014635 InChI Key: CFOJQUGXHMGMOT-UHFFFAOYSA-N Synonym: 3-pyridylthiourea,1-3-pyridyl-2-thiourea,1-pyridin-3-yl thiourea,pyridin-3-yl-thiourea,n-3-pyridyl thiourea,n-pyridin-3-ylthiourea,pyridin-3-yl thiourea,1-pyridin-3-ylthiourea,amino 3-pyridylamino methane-1-thione,3-pyridinylthiourea PubChem CID: 2760515 IUPAC Name: pyridin-3-ylthiourea SMILES: NC(=S)NC1=CC=CN=C1