accessibility menu, dialog, popup

UserName

CAS RN 30097-06-4

IUPAC Name: (2S,4S)-1,3-thiazolidin-3-ium-2,4-dicarboxylate
Synonyms: tidiacic
Molecular Weight (g/mol): 176.17
Molecular Formula: C5H6NO4S
InChi Key: DAXBISKSIDBYEU-GBXIJSLDSA-M
SMILES: [O-]C(=O)[C@H]1CS[C@H]([NH2+]1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results