CAS RN 300816-22-2
IUPAC Name:
12-chloro-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraene
Synonyms:
12-chloro-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(12),2(6),8,10-tetraene
Molecular Weight (g/mol):
210.68
Molecular Formula:
C9H7ClN2S
InChi Key:
JKXJMJPJOYMQSK-UHFFFAOYSA-N
SMILES:
ClC1=C2C3=C(CCC3)SC2=NC=N1
1
–
1
of
1
results
1
Promotions Available
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine, 96%
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine, 96%
CAS: 300816-22-2 Molecular Formula: C9H7ClN2S Molecular Weight (g/mol): 210.68 MDL Number: MFCD00572400 InChI Key: JKXJMJPJOYMQSK-UHFFFAOYSA-N PubChem CID: 789817 IUPAC Name: 1-chloro-7,8-dihydro-6H-cyclopenta[2,3]thieno[2,4-d]pyrimidine SMILES: ClC1=C2C3=C(CCC3)SC2=NC=N1