CAS RN 299442-43-6
IUPAC Name:
2-amino-7-(furan-2-yl)-5,6,7,8-tetrahydroquinazolin-5-one
Synonyms:
2-amino-7-(furan-2-yl)-7,8-dihydro-6H-quinazolin-5-one
Molecular Weight (g/mol):
229.24
Molecular Formula:
C12H11N3O2
InChi Key:
SOJUSNIBPPMLCC-UHFFFAOYNA-N
SMILES:
NC1=NC=C2C(=O)CC(CC2=N1)C1=CC=CO1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Adenylyl Cyclase Type V Inhibitor, NKY80
CAS: 299442-43-6 Molecular Formula: C12H11N3O2,C12H11N3O2 Molecular Weight (g/mol): 229.24 InChI Key: SOJUSNIBPPMLCC-UHFFFAOYNA-N Synonym: NKY80; 2-Amino-7-(furan-2-yl)-7,8-dihydroquinazolin-5(6H)-one IUPAC Name: 2-amino-7-(furan-2-yl)-5,6,7,8-tetrahydroquinazolin-5-one SMILES: NC1=NC=C2C(=O)CC(CC2=N1)C1=CC=CO1
2
Tocris Bioscience™ NKY 80
CAS: 299442-43-6 Molecular Formula: C12H11N3O2