missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenylyl Cyclase Type V Inhibitor, NKY80

Catalog No. AAJ64042LB0
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64042LB0 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64042LB0 Supplier Thermo Scientific Chemicals Supplier No. J64042LB0
Only null left

 

A cell-permeable quinazolinone containing a non-nucleoside compound that acts as a potent, selective inhibitor of adenylyl cyclase (AC) type V isoform

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 299442-43-6
Molecular Formula C12H11N3O2
Molecular Weight (g/mol) 229.24
InChI Key SOJUSNIBPPMLCC-UHFFFAOYNA-N
Synonym NKY80; 2-Amino-7-(furan-2-yl)-7,8-dihydroquinazolin-5(6H)-one
IUPAC Name 2-amino-7-(furan-2-yl)-5,6,7,8-tetrahydroquinazolin-5-one
SMILES NC1=NC=C2C(=O)CC(CC2=N1)C1=CC=CO1

Specifications

Quantity 5 mg
Description Pale orange
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable quinazolinone containing a non-nucleoside compound that acts as a potent, selective inhibitor of adenylyl cyclase (AC) type V isoform
Formula Weight 229.23
Sensitivity Light Sensitive
Solubility Soluble in DMSO or methanol
Product Type Adenylyl Cyclase Type V Inhibitor
Form Powder
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only