accessibility menu, dialog, popup

UserName

CAS RN 29934-17-6

IUPAC Name: dichloropalladium; bis(tricyclohexylphosphane)
Synonyms: palladium chloride; bis(tricyclohexylphosphine)
Molecular Weight (g/mol): 738.19
Molecular Formula: C36H66Cl2P2Pd
InChi Key: VUYVXCJTTQJVKJ-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Physical Form 
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)