missing translation for 'onlineSavingsMsg'
Learn More

Dichlorobis(tricyclohexylphosphine)palladium(II), 98%

Catalog No. AC395892500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395892500 250 mg Glass bottle
AC395890010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC395892500 Supplier Thermo Scientific Chemicals Supplier No. 395892500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 29934-17-6 | C36H66Cl2P2Pd | 738.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29934-17-6
Molecular Formula C36H66Cl2P2Pd
Molecular Weight (g/mol) 738.19
MDL Number MFCD00191830
InChI Key VUYVXCJTTQJVKJ-UHFFFAOYSA-L
Synonym dichlorobis tricyclohexylphosphine palladium ii, bis tricyclohexylphosphine palladium ii dichloride, dichlorobis tricyclohexylphosphine palladium, bis tricyclohexylphosphine-palladium ii dichloride, bis tricyclohexylphosphine palladium ii-dichloride, bis tricyclo-hexylphosphine palladium ii dichloride, bis tricyclohexylphosphine palladium ii chloride
PubChem CID 11050900
SMILES Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 270°C
Color Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
IUPAC Name dichloropalladium; bis(tricyclohexylphosphane)
Formula Weight 738.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dichlorobis(tricyclohexylphosphine)palladium(II)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only