missing translation for 'onlineSavingsMsg'
Learn More

Bis(tricyclohexylphosphine)palladium(II) Dichloride 97.0+%, TCI America™

Catalog No. B20555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20555G 5 g
B20551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20555G Supplier TCI America Supplier No. B20555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 29934-17-6
Molecular Formula C36H66Cl2P2Pd
Molecular Weight (g/mol) 738.19
MDL Number MFCD00191830
InChI Key VUYVXCJTTQJVKJ-UHFFFAOYSA-L
Synonym dichlorobis tricyclohexylphosphine palladium ii, bis tricyclohexylphosphine palladium ii dichloride, dichlorobis tricyclohexylphosphine palladium, bis tricyclohexylphosphine-palladium ii dichloride, bis tricyclohexylphosphine palladium ii-dichloride, bis tricyclo-hexylphosphine palladium ii dichloride, bis tricyclohexylphosphine palladium ii chloride
PubChem CID 11050900
IUPAC Name dichloropalladium; bis(tricyclohexylphosphane)
SMILES Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 738.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(tricyclohexylphosphine)palladium(II) Dichloride
TSCA No
Recommended Storage Freezer