accessibility menu, dialog, popup

UserName

CAS RN 29851-57-8

IUPAC Name: 6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-8,9-dihydro-7H-purin-8-one
Synonyms: 6-amino-9-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-7H-purin-8-one
Molecular Weight (g/mol): 283.24
Molecular Formula: C10H13N5O5
InChi Key: UEHOMUNTZPIBIL-UHFFFAOYNA-N
SMILES: NC1=C2NC(=O)N(C3OC(CO)C(O)C3O)C2=NC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results