accessibility menu, dialog, popup

UserName

CAS RN 29683-23-6

IUPAC Name: thian-4-ol
Synonyms: thian-4-ol
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: YODQQARABJQLIP-UHFFFAOYSA-N
SMILES: OC1CCSCC1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)