accessibility menu, dialog, popup

UserName

CAS RN 29681-43-4

H3CONOOCH3

CAS RN 29681-43-4

IUPAC Name: methyl 4-methoxypyridine-2-carboxylate
Synonyms: methyl 4-methoxypyridine-2-carboxylate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: OJDKENGKKYVJLY-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC=CC(OC)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)