CAS RN 29681-42-3
CAS RN 29681-42-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 4-bromopyridine-2-carboxylate, 97+%
CAS: 29681-42-3 Molecular Formula: C7H6BrNO2 Molecular Weight (g/mol): 216.034 MDL Number: MFCD07374897 InChI Key: JZFLATQBIPILFS-UHFFFAOYSA-N PubChem CID: 12828947 IUPAC Name: methyl 4-bromopyridine-2-carboxylate SMILES: COC(=O)C1=NC=CC(=C1)Br