accessibility menu, dialog, popup

UserName

CAS RN 29658-97-7

H3COHOOHO(E)

CAS RN 29658-97-7

IUPAC Name: 2-[(1E)-dodec-1-en-1-yl]butanedioic acid
Synonyms: 2-[(1E)-dodec-1-en-1-yl]butanedioic acid
Molecular Weight (g/mol): 284.4
Molecular Formula: C16H28O4
InChi Key: QDCPNGVVOWVKJG-VAWYXSNFNA-N
SMILES: CCCCCCCCCC\C=C\C(CC(O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results