CAS RN 2964-48-9
IUPAC Name:
(1S,2S)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Synonyms:
(1S,2S)-1,3-dihydroxy-1-(4-nitrophenyl)propan-2-aminium
Molecular Weight (g/mol):
213.21
Molecular Formula:
C9H13N2O4
InChi Key:
OCYJXSUPZMNXEN-IUCAKERBSA-O
SMILES:
[NH3+][C@@H](CO)[C@@H](O)C1=CC=C(C=C1)[N+]([O-])=O
1
–
2
of
2
results
2
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific™
CAS: 2964-48-9 Molecular Formula: C9H12N2O4 Molecular Weight (g/mol): 212.2 MDL Number: MFCD00007359 InChI Key: OCYJXSUPZMNXEN-IUCAKERBSA-N Synonym: 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol,1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol,1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol,2-amino-1-4-nitro-phenyl-propane-1,3-diol,1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s,threomine,1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol,1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned,1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s,l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol PubChem CID: 224171 SMILES: C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]