missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific™

Catalog No. p-3776750
Change view
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC334800050 5g Glass bottle
AC334800250 25g Glass bottle
AC334801000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC334800050 Supplier Thermo Scientific Chemicals Supplier No. 334800050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2964-48-9
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.2
MDL Number MFCD00007359
InChI Key OCYJXSUPZMNXEN-IUCAKERBSA-N
Synonym 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol, 1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol, 1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol, 2-amino-1-4-nitro-phenyl-propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s, threomine, 1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol, 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned, 1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s, l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol
PubChem CID 224171
SMILES C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 5g
Specific Rotation + 31
Formula Weight 212.2
Percent Purity 96%
Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
EINECSNumber 221-001-4

RUO – Research Use Only