missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-(4-nitrophenyl)propane-1,3-diol, MilliporeSigma™ Supelco™

Catalog No. 111002190
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002190 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002190 Supplier Merck Emd Millipore Supplier No. BP417

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 2964-48-9
Linear Formula O2 NC6H4 CH(OH)CH(NH2)CH2 OH
MDL Number MFCD00007359
Melting Point 163°C to 166°C
Synonym (1 S,2 S)-(+)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol; L-(+)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 212.20
Grade Pharmaceutical Primary Standard