accessibility menu, dialog, popup

UserName

CH3SSOOH3C

CAS RN 2943-42-2

IUPAC Name: 1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene
Synonyms: 1-methyl-4-[(4-methylbenzenesulfonyl)sulfanyl]benzene
Molecular Weight (g/mol): 278.38
Molecular Formula: C14H14O2S2
InChi Key: HSAROCRPZOYKGS-UHFFFAOYSA-N
SMILES: CC1=CC=C(SS(=O)(=O)C2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results