accessibility menu, dialog, popup

UserName

FFFSOOONa

CAS RN 2926-30-9

IUPAC Name: sodium trifluoromethanesulfonate
Synonyms: sodium triflate
Molecular Weight (g/mol): 172.05
Molecular Formula: CF3NaO3S
InChi Key: XGPOMXSYOKFBHS-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (2)
  • (5)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (2)
  • (8)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)