missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mequitazine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 148°C |
| Color | White |
| Quantity | 1 g |
| UN Number | 2811 |
| Formula Weight | 322.47 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Mequitazine |
Safety and Handling
| RTECSNumber | SP5080000 |
| TSCA | No |