accessibility menu, dialog, popup

UserName

N

CAS RN 2920-38-9

IUPAC Name: [1,1'-biphenyl]-4-carbonitrile
Synonyms: [1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 179.22
Molecular Formula: C13H9N
InChi Key: BPMBNLJJRKCCRT-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)