missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobiphenyl, 95%

Catalog No. AAH2679118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2679118 50 g
AAH2679109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2679118 Supplier Thermo Scientific Chemicals Supplier No. H2679118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2920-38-9 | C13H9N | 179.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2920-38-9
Molecular Formula C13H9N
Molecular Weight (g/mol) 179.22
MDL Number MFCD00001821
InChI Key BPMBNLJJRKCCRT-UHFFFAOYSA-N
Synonym 4-cyanobiphenyl, 1,1'-biphenyl-4-carbonitrile, 4-biphenylcarbonitrile, p-phenylbenzonitrile, p-cyanobiphenyl, biphenyl-4-carbonitrile, 4-phenylbenzenecarbonitrile, 4-biphenylcarboxylic acid nitrile, 4-cyanodiphenyl
PubChem CID 18021
IUPAC Name 4-phenylbenzonitrile
SMILES N#CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 85°C to 87°C
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN3439
Beilstein 1907924
Formula Weight 179.22
Percent Purity 95%
Chemical Name or Material 4-Cyanobiphenyl
Hazard Category H302+H312+H332-H315-H319-H335-H500
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 220-860-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only