accessibility menu, dialog, popup

UserName

CAS RN 2903-48-2

IUPAC Name: [(2,6-dimethylphenyl)carbamoyl]formic acid
Synonyms: [(2,6-dimethylphenyl)carbamoyl]formic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: JFTAPSBIODPQJN-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1NC(=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results