accessibility menu, dialog, popup

UserName

CAS RN 29022-11-5

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetic acid
Synonyms: {[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetic acid
Molecular Weight (g/mol): 297.31
Molecular Formula: C17H15NO4
InChi Key: NDKDFTQNXLHCGO-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)