CAS RN 2894848-07-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ BI 3231
Potent and selective hydroxysteroid 17β dehydrogenase 13 (HSD17B13) inhibitor