accessibility menu, dialog, popup

UserName

H3COOCH3OO

CAS RN 2892-62-8

IUPAC Name: dibutoxycyclobut-3-ene-1,2-dione
Synonyms: squaric acid dibutyl ester sadbe
Molecular Weight (g/mol): 226.27
Molecular Formula: C12H18O4
InChi Key: XBRWELTXMQSEIN-UHFFFAOYSA-N
SMILES: CCCCOC1=C(OCCCC)C(=O)C1=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)