accessibility menu, dialog, popup

UserName

CAS RN 288-13-1

IUPAC Name: 1H-pyrazole
Synonyms: pyrazole pyrazole (substance) Hpz
Molecular Weight (g/mol): 68.08
Molecular Formula: C3H4N2
InChi Key: WTKZEGDFNFYCGP-UHFFFAOYSA-N
SMILES: N1C=CC=N1
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)