accessibility menu, dialog, popup

UserName

NOHCClClNOHCClClHClPdClHPd(E/Z)(E/Z)

CAS RN 287410-78-0

IUPAC Name: (chloraniumylpalladio)chloranium bis(5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]benzen-1-ide) palladium
Synonyms: (chloraniumylpalladio)chloranium bis(5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]benzen-1-ide) palladium
Molecular Weight (g/mol): 815.98
Molecular Formula: C26H18Cl6N2O2Pd2
InChi Key: QPUPWYGEQHWYOF-UHFFFAOYSA-N
SMILES: [Pd].[ClH+][Pd][ClH+].ON=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=[C-]1.ON=C(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=[C-]1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results