missing translation for 'onlineSavingsMsg'
Learn More

Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) Dimer 95.0+%, TCI America™

Catalog No. D3806250MG
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3806250MG 250 mg
D38061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3806250MG Supplier TCI America Supplier No. D3806250MG
Only null left

Chemical Identifiers

CAS 287410-78-0
Molecular Formula C26H18Cl6N2O2Pd2
Molecular Weight (g/mol) 815.982
MDL Number MFCD09038424
InChI Key QPUPWYGEQHWYOF-INIQRQEMSA-N
Synonym Najera Catalyst I
PubChem CID 132774960
IUPAC Name (NE)-N-[(4-chlorobenzene-6-id-1-yl)-(4-chlorophenyl)methylidene]hydroxylamine;dichloroniopalladium;palladium
SMILES C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.C1=CC(=CC=C1C(=NO)C2=CC=C(C=[C-]2)Cl)Cl.[ClH+][Pd][ClH+].[Pd]

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 813.97
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Di-mu-chlorobis[5-chloro-2-[(4-chlorophenyl)(hydroxyimino)methyl]phenyl]palladium(II) Dimer