CAS RN 2873-97-4
IUPAC Name:
N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide
Synonyms:
diacetone acrylamide
Molecular Weight (g/mol):
169.22
Molecular Formula:
C9H15NO2
InChi Key:
OMNKZBIFPJNNIO-UHFFFAOYSA-N
SMILES:
CC(=O)CC(C)(C)NC(=O)C=C
1
–
1
of
1
results
1
Promotions Available
Diacetone acrylamide, 99%
Diacetone acrylamide, 99%
CAS: 2873-97-4 Molecular Formula: C9H15NO2 Molecular Weight (g/mol): 169.22 MDL Number: MFCD00008788 InChI Key: OMNKZBIFPJNNIO-UHFFFAOYSA-N Synonym: diacetone acrylamide,diacetoneacrylamide,n-1,1-dimethyl-3-oxobutyl acrylamide,2-propenamide, n-1,1-dimethyl-3-oxobutyl,n-2-methyl-4-oxopentan-2-yl acrylamide,n-2-2-methyl-4-oxopentyl acrylamide,acrylamide, n-1,1-dimethyl-3-oxobutyl,n-1,1-dimethyl-3-oxobutyl-2-propenamide,acrylamide, n,n-diacetonyl,ccris 5898 PubChem CID: 17888 IUPAC Name: N-(2-methyl-4-oxopentan-2-yl)prop-2-enamide SMILES: CC(=O)CC(C)(C)NC(=O)C=C