accessibility menu, dialog, popup

UserName

CAS RN 2871-01-4

NH2HNOHNOO

CAS RN 2871-01-4

IUPAC Name: 2-[(4-amino-2-nitrophenyl)amino]ethan-1-ol
Synonyms: HC red NO. 3 HC red no. 3
Molecular Weight (g/mol): 197.19
Molecular Formula: C8H11N3O3
InChi Key: GZGZVOLBULPDFD-UHFFFAOYSA-N
SMILES: NC1=CC=C(NCCO)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results