accessibility menu, dialog, popup

UserName

CAS RN 28667-63-2

SS

CAS RN 28667-63-2

IUPAC Name: 3,10-dithiatricyclo[10.2.2.2⁵,⁸]octadeca-1(14),5,7,12,15,17-hexaene
Synonyms: 3,10-dithiatricyclo[10.2.2.2⁵,⁸]octadeca-1(14),5,7,12,15,17-hexaene
Molecular Weight (g/mol): 272.42
Molecular Formula: C16H16S2
InChi Key: CUWSXVJIPZWUGC-UHFFFAOYSA-N
SMILES: C1SCC2=CC=C(CSCC3=CC=C1C=C3)C=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results