accessibility menu, dialog, popup

UserName

CAS RN 28553-12-0

H3CCH3OOOOCH3CH3

CAS RN 28553-12-0

IUPAC Name: 1,2-bis(7-methyloctyl) benzene-1,2-dicarboxylate
Synonyms: diisononyl phthalate DINP jayflex dinp jay-dinp palatinol N phthalisocizer dinp witamol 150 di-isononylphthalate vinylcizer 90 sansocizer dinp
Molecular Weight (g/mol): 418.62
Molecular Formula: C26H42O4
InChi Key: HBGGXOJOCNVPFY-UHFFFAOYSA-N
SMILES: CC(C)CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCC(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)