accessibility menu, dialog, popup

UserName

CH3NOHNFFF

CAS RN 2836-44-4

IUPAC Name: 2-methyl-6-(trifluoromethyl)-1,4-dihydropyrimidin-4-one
Synonyms: 2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
Molecular Weight (g/mol): 178.11
Molecular Formula: C6H5F3N2O
InChi Key: DBDRDWKBAGNHHS-UHFFFAOYSA-N
SMILES: CC1=NC(=O)C=C(N1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results