accessibility menu, dialog, popup

UserName

CAS RN 28286-88-6

IUPAC Name: 2-(2,6-dimethyl-4H-pyran-4-ylidene)propanedinitrile
Synonyms: 2-(2,6-dimethylpyran-4-ylidene)propanedinitrile
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: XYBUCJYJVULPHW-UHFFFAOYSA-N
SMILES: CC1=CC(C=C(C)O1)=C(C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results