accessibility menu, dialog, popup

UserName

NH3(R)(R)HO

CAS RN 28250-45-5

IUPAC Name: (1R,2R)-2-(hydroxymethyl)cyclohexan-1-aminium
Synonyms: (1R,2R)-2-(hydroxymethyl)cyclohexan-1-aminium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: GCWPGEWXYDEQAY-NKWVEPMBSA-O
SMILES: [NH3+][C@@H]1CCCC[C@H]1CO
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)