accessibility menu, dialog, popup

UserName

CAS RN 2810-04-0

H3COOS

CAS RN 2810-04-0

IUPAC Name: ethyl thiophene-2-carboxylate
Synonyms: thiophene, 2-ethyl carboxylate
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: JZGZKRJVTIRPOK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CS1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)