accessibility menu, dialog, popup

UserName

CAS RN 28020-73-7

HNNNNHN

CAS RN 28020-73-7

IUPAC Name: 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole
Synonyms: 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole
Molecular Weight (g/mol): 311.35
Molecular Formula: C19H13N5
InChi Key: JBKICBDXAZNSKA-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2N=C1C1=CC=CC(=N1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results