accessibility menu, dialog, popup

UserName

CAS RN 279-23-2

IUPAC Name: bicyclo[2.2.1]heptane
Synonyms: norbornane
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: UMRZSTCPUPJPOJ-UHFFFAOYSA-N
SMILES: C1CC2CCC1C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results