accessibility menu, dialog, popup

UserName

CAS RN 27724-96-5

IUPAC Name: hydrogen 3-{4-[4-(aminomethyl)cyclohexanecarbonyloxy]phenyl}propanoic acid chloride
Synonyms: hydrogen cetraxate chloride
Molecular Weight (g/mol): 341.83
Molecular Formula: C17H24ClNO4
InChi Key: USROQQUKEBHOFF-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1CCC(CC1)C(=O)OC1=CC=C(CCC(O)=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results