accessibility menu, dialog, popup

UserName

BrONOP

CAS RN 2767-70-6

IUPAC Name: [(4-nitrophenyl)methyl]triphenylphosphanium bromide
Synonyms: [(4-nitrophenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 478.33
Molecular Formula: C25H21BrNO2P
InChi Key: IPJPTPFIJLFWLP-UHFFFAOYSA-M
SMILES: [Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results