missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrobenzyl)triphenylphosphonium bromide, 98%

Catalog No. AAA1528814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1528814 25 g
AAA1528806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1528814 Supplier Thermo Scientific Chemicals Supplier No. A1528814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2767-70-6 | C25H21BrNO2P | 478.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2767-70-6
Molecular Formula C25H21BrNO2P
Molecular Weight (g/mol) 478.33
MDL Number MFCD00032108
InChI Key IPJPTPFIJLFWLP-UHFFFAOYSA-M
Synonym 4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide
PubChem CID 197008
IUPAC Name (4-nitrophenyl)methyl-triphenylphosphanium;bromide
SMILES [Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point ∼280°C (decomposition)
Quantity 25 g
Formula Weight 478.33
Percent Purity 98%
Chemical Name or Material (4-Nitrobenzyl)triphenylphosphonium bromide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-444-0
RTECSNumber TA2390000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only