missing translation for 'onlineSavingsMsg'
Learn More

(4-Nitrobenzyl)triphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. N070125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N070125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N070125G Supplier TCI America Supplier No. N070125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2767-70-6
Molecular Formula C25H21BrNO2P
Molecular Weight (g/mol) 478.33
MDL Number MFCD00032108
InChI Key IPJPTPFIJLFWLP-UHFFFAOYSA-M
Synonym 4-nitrobenzyl triphenylphosphonium bromide, p-nitrobenzyl triphenylphosphonium bromide, phosphonium, p-nitrobenzyl triphenyl-, bromide, 4-nitrobenzyl triphenylphosphoniumbromide, 4-nitrobenzyl triphenyl phosphonium bromide, 4-nitrophenyl methyl triphenylphosphanium bromide, acmc-209gym, phosphonium, 4-nitrophenyl methyl triphenyl-, bromide, wln: wnr d1pr&r&r &e, 4-nitrobenzyltriphenylphosphonium bromide
PubChem CID 197008
IUPAC Name [(4-nitrophenyl)methyl]triphenylphosphanium bromide
SMILES [Br-].[O-][N+](=O)C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 275°C
Color White-Yellow
Quantity 25 g
Formula Weight 478.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Nitrobenzyl)triphenylphosphonium Bromide
RTECSNumber TA2390000
TSCA No