accessibility menu, dialog, popup

UserName

MoOHHOOO

CAS RN 27546-07-2

IUPAC Name: dioxomolybdenumdiol
Synonyms: molybdic acid H2MoO4 molybdic(VI) acid
Molecular Weight (g/mol): 161.96
Molecular Formula: H2MoO4
InChi Key: VLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILES: O[Mo](O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results